Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Hydrogen hexafluoroantimonate(V), ≈65% aq. soln.
Click to view available options
Quantity:
25g
50g
100g
Specifications
Concentration or Composition (by Analyte or Components) | ≈65% aq. soln. |
Quantity | 100g |
Color | Colorless to Orange |
Linear Formula | HF·SbF5 |
UN Number | UN3264 |
Solubility Information | It is fully miscible with water,soluble in SO2ClF,SO2. |
Formula Weight | 236.75 |
Physical Form | Liquid |
Chemical Name or Material | Hydrogen hexafluoroantimonate(V) |
Safety and Handling
GHS H Statement
H314-H302-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 241-023-8
TSCA : Yes
Recommended Storage : Ambient temperatures