Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 17.5% w/w, Reagents

Hydrogen Peroxide, 17.5% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CH300000200EV

Quantity 200 L

Catalog No. 01-334-974


Only null left
Add to Cart

Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 7722-84-1
18.02
Dihydrogen Dioxide, Oxydol
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

Colorless
105°C
Dihydrogen Dioxide, Oxydol
O
18.02
Reagent
1.0629
200 L
Liquid
7732-18-5 , 7722-84-1
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
17.5%
Poly Drum
2 to 4.8
Non-sterile
Hydrogen Peroxide, 17.5% w/w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.