Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrogen Peroxide, 50% w/w, Reagents
Hydrogen Peroxide, 50% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc C2214680500LB
Quantity | 500 lb. |
---|
Chemical Identifiers
7732-18-5, 7722-84-1 | |
18.02 | |
Dihydrogen Dioxide, Oxydol | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
Colorless | |
126°C | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Poly Drum | |
<3 | |
Non-sterile | |
Hydrogen Peroxide, 50% w/w |
7732-18-5, 7722-84-1 | |
-52°C | |
Dihydrogen Dioxide, Oxydol | |
O | |
18.02 | |
1.19 | |
500 lb. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction