Learn More
Aat BioquestSupplier Diversity Partner ReadiUse™ hydrogen peroxide solution *50 mM calibrated and stabilized solution*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 11004
Compared to other commercial hydrogen peroxide solutions, our formulated hydrogen peroxide solution is much more stable. It is calibrated to ensure more reproducible peroxidase-based assays.
Catalog No. 50-195-3329
Due to product restrictions, please to purchase or view availability for this product.