Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide Solution AR 30%, Macron Fine Chemicals™ ™

Click to view available options
Quantity:
500mL
4L
Packaging:
Poly
7722-84-1
H2O2
34.014
MHAJPDPJQMAIIY-UHFFFAOYSA-N
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
784
CHEBI:16240
hydrogen peroxide
OO
7722-84-1
H2O2
34.014
MHAJPDPJQMAIIY-UHFFFAOYSA-N
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
784
CHEBI:16240
hydrogen peroxide
OO
Assay Percent Range 30%
Formula Weight 34.01
Packaging Poly
Quantity 4L
Chemical Name or Material Hydrogen Peroxide Solution