Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative, 2.5 to 3.5%, U.S.P., J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

7722-84-1
H2O2
34.014
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
CHEBI:16240
Specific Gravity 1L = 1.00kg
Quantity 4L
CAS 7722-84-1
Molecular Formula H2O2
Packaging Narrow Mouth Amber Glass Bottle
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name hydrogen peroxide
Molecular Weight (g/mol) 34.014
PubChem CID 784
ChEBI CHEBI:16240
Formula Weight 34.01
Chemical Name or Material Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative
Assay 2.5 to 3.5%
Grade USP
Show More Show Less