Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide Topical Solution, USP, 2.5-3.5%, Spectrum™

Click to view available options
Quantity:
16 oz.
1 gal.
6 x 16 oz.
4 x 1 gal.
200 L
Packaging:
Poly Bottle
Poly Drum
7722-84-1
H2O2
34.01
MHAJPDPJQMAIIY-UHFFFAOYSA-N
peroxol
OO
7722-84-1
H2O2
34.01
MHAJPDPJQMAIIY-UHFFFAOYSA-N
peroxol
OO

HY110, 7722-84-1, H2O2

Spectrum™ Hydrogen Peroxide Topical Solution, USP is a mild antiseptic used on the skin to prevent infection of minor cuts, scrapes, and burns. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 2.5 to 3.5%
Non_volatile Matter 0.0015
Packaging Poly Bottle
Quantity 4 x 1 gal.