Alfa Aesar™ Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min

Manufacturer: Alfa Aesar™ 1232506

Quantity 5g

 View more versions of this product

Catalog No. AA1232506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 27988-77-8
    Formula Weight 339.79 (Anhydrous)
    MDL Number MFCD00149903
    UN Number UN3260
    Synonym Tetrachloroauric(III) acid; Chloroauric acid
    Chemical Name or Material Hydrogen tetrachloroaurate(III) hydrate

    Specifications

    Formula Weight 339.79 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00149903
    Health Hazard 2 GHS H Statement
    H314-H318-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    TSCA TSCA
    Physical Form Crystalline
    CAS 27988-77-8
    UN Number UN3260
    Synonym Tetrachloroauric(III) acid; Chloroauric acid
    Health Hazard 1 Danger
    Sensitivity Light Sensitive & Hygroscopic
    Color Yellow-orange
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    RTECS Number MD5428000
    Concentration Au 49% min.
    Quantity 5g
    Chemical Name or Material Hydrogen tetrachloroaurate(III) hydrate
    Assay 99.9%, (metals basis)

    GHS H Statement
    H314-H318-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit