Alfa Aesar™ Hydrogen tetranitratoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 38.9-40.1%

Manufacturer: Alfa Aesar™ 4043103

Quantity 1g

 View more versions of this product

Catalog No. AA4043103

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 445.99 (Anhydrous)
    MDL Number MFCD00046178
    UN Number UN1477
    Synonym Nitratoauric acid
    Chemical Name or Material Hydrogen tetranitratoaurate(III) hydrate
    Grade Premion™

    Specifications

    Formula Weight 445.99 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Decomposes in water; Soluble in HNO3
    Health Hazard 2 GHS H Statement
    H272-H315-H319
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    Physical Form Crystalline
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
    MDL Number MFCD00046178
    UN Number UN1477
    Synonym Nitratoauric acid
    Melting Point ≈72°C (dec.)
    Health Hazard 1 Danger
    Quantity 1g
    Concentration Au 38.9 to 40.1%
    Chemical Name or Material Hydrogen tetranitratoaurate(III) hydrate
    Assay 99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H272-H315-H319
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit