Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinidine Hydrochloride 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
1476-98-8
C20H27ClN2O2
362.898
MFCD00135598
MULXTQKDWYBJMO-VJAUXQICSA-N
Dihydroquinidine Hydrochloride
16211709
(S)-[(2R,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;hydrochloride
CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OC)O.Cl
1476-98-8
C20H27ClN2O2
362.898
MFCD00135598
MULXTQKDWYBJMO-VJAUXQICSA-N
Dihydroquinidine Hydrochloride
16211709
(S)-[(2R,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;hydrochloride
CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OC)O.Cl
Color White
Quantity 250g
Formula Weight 362.90
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T,HPLC)
Chemical Name or Material Hydroquinidine Hydrochloride

RTECSNumber : MX3018000

TSCA : No