Alfa Aesar™ Hydroquinone bis(2-hydroxyethyl) ether, 95%

Manufacturer: Alfa Aesar™ B2403722

Quantity 100g

 View more versions of this product

Catalog No. AAB2403722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 104-38-1
    Molecular Formula C10H14O4
    Formula Weight 198.22
    MDL Number MFCD00002861
    Synonym 1,4-Bis(2-hydroxyethoxy)benzene
    Chemical Name or Material Hydroquinone bis(2-hydroxyethyl) ether

    Specifications

    Formula Weight 198.22
    CAS 104-38-1
    Melting Point 100° to 102°C
    Quantity 100g
    TSCA TSCA
    Molecular Formula C10H14O4
    MDL Number MFCD00002861
    Synonym 1,4-Bis(2-hydroxyethoxy)benzene
    Chemical Name or Material Hydroquinone bis(2-hydroxyethyl) ether
    Assay 95%

    Description

    Applications
    Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. Protect from humidity and water. It is hygroscopic in nature. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit