Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinone-O,O'-diacetic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
2245-53-6
C10H10O6
226.184
MFCD00016816
DNXOCFKTVLHUMU-UHFFFAOYSA-N
2,2'-1,4-phenylenebis oxy diacetic acid,1,4-phenylenedioxydiacetic acid,hydroquinone-o,o'-diacetic acid,1,4-dicarboxymethoxybenzene,hydroquinone-o,o-diacetic acid,acetic acid, p-phenylenedioxy di,acetic acid, 2,2'-1,4-phenylenebis oxy bis,2-4-carboxymethoxy phenoxy acetic acid
75251
2-[4-(carboxymethoxy)phenoxy]acetic acid
C1=CC(=CC=C1OCC(=O)O)OCC(=O)O
2245-53-6
C10H10O6
226.184
MFCD00016816
DNXOCFKTVLHUMU-UHFFFAOYSA-N
2,2'-1,4-phenylenebis oxy diacetic acid,1,4-phenylenedioxydiacetic acid,hydroquinone-o,o'-diacetic acid,1,4-dicarboxymethoxybenzene,hydroquinone-o,o-diacetic acid,acetic acid, p-phenylenedioxy di,acetic acid, 2,2'-1,4-phenylenebis oxy bis,2-4-carboxymethoxy phenoxy acetic acid
75251
2-[4-(carboxymethoxy)phenoxy]acetic acid
C1=CC(=CC=C1OCC(=O)O)OCC(=O)O
Quantity 25g
Color White to Tan
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 95%
Linear Formula C6H4(OCH2COOH)2
Beilstein 06, IV, 5746
Formula Weight 226.19
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material Hydroquinone-O, O';-diacetic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 218-834-0

TSCA : TSCA