Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hydroquinone-O,O'-diacetic acid, 98%

Click to view available options
Quantity:
5g
25g
100g
2245-53-6
C10H10O6
226.184
MFCD00016816
DNXOCFKTVLHUMU-UHFFFAOYSA-N
2,2'-1,4-phenylenebis oxy diacetic acid,1,4-phenylenedioxydiacetic acid,hydroquinone-o,o'-diacetic acid,1,4-dicarboxymethoxybenzene,hydroquinone-o,o-diacetic acid,acetic acid, p-phenylenedioxy di,acetic acid, 2,2'-1,4-phenylenebis oxy bis,2-4-carboxymethoxy phenoxy acetic acid
75251
2-[4-(carboxymethoxy)phenoxy]acetic acid
C1=CC(=CC=C1OCC(=O)O)OCC(=O)O
2245-53-6
C10H10O6
226.184
MFCD00016816
DNXOCFKTVLHUMU-UHFFFAOYSA-N
2,2'-1,4-phenylenebis oxy diacetic acid,1,4-phenylenedioxydiacetic acid,hydroquinone-o,o'-diacetic acid,1,4-dicarboxymethoxybenzene,hydroquinone-o,o-diacetic acid,acetic acid, p-phenylenedioxy di,acetic acid, 2,2'-1,4-phenylenebis oxy bis,2-4-carboxymethoxy phenoxy acetic acid
75251
2-[4-(carboxymethoxy)phenoxy]acetic acid
C1=CC(=CC=C1OCC(=O)O)OCC(=O)O
Applications
Hydroquinone-O,O′-diacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Melting Point ~255°C (decomposition)
Quantity 25g
Beilstein 1984454
Formula Weight 226.19
Percent Purity 98%
Chemical Name or Material Hydroquinone-O,O'-diacetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA