Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinone, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
123-31-9
C6H6O2
110.11
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
benzene-1,4-diol
OC1=CC=C(O)C=C1
123-31-9
C6H6O2
110.11
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
benzene-1,4-diol
OC1=CC=C(O)C=C1

HY113, 123-31-9, C6H6O2

Spectrum™ Hydroquinone, USP is an effective topical medicine used to treat dark colored patches, freckles, and spots on the skin that appear due to sun exposure. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Melting Range 172°-174°C
Ignition Residue 0.005
Quantity 25 kg
CAS Max % 1
UN Number UN3077
Percent Purity 99 to 100.5%