Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HydroSpec™ Coulometric CG, Ricca Chemical
Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.
$104.00
Specifications
Specific Gravity | 0.93 |
---|---|
CAS | 111-90-0, 67-56-1, 288‐32‐4, 7446‐09‐5, 68007-08-9 |
CAS Min % | 60, 10, 5, 5, 5 |
CAS Max % | 90, 30, 20, 20, 10 |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
RK130000110
|
Ricca Chemical Company
RK130000110C |
110 mL |
Each of 1 for $104.00
|
|
|||||
Specifications
0.93 | |
60, 10, 5, 5, 5 | |
Liquid |
111-90-0, 67-56-1, 288‐32‐4, 7446‐09‐5, 68007-08-9 | |
90, 30, 20, 20, 10 | |
UN1230 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title