Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrospec™ Titrant K5 for Two Component Volumetric Titrations of Aldehydes and Ketones, Ricca Chemical PROMO

Achieve high performance and reliable results. Hydrospec™ supports a wide range of sample types that help you produce the fast, reliable, and reproducible results you are looking for.

Manufacturer:  Ricca Chemical Company RK3000006X500CPM

 View more versions of this product

Catalog No. 00-000-6500PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Description

Description

  • Reliable, Safe, Pyridine-Free
  • Available in 500mL and 1L sizes
  • 1L Bottles have the Industry Standard GL45 Neck Finish for Convenient Connection
  • Stable Formulations and Comprehensive Chemical Compatibility
  • 5 Year Shelf Life for Most Reagents
  • Technical Support Every Step of the Way
  • Tight Specifications for More Accurate Results and Low Lot-to-Lot Variability
  • Serving Scientists Successfully for Over 45 Years
Specifications

Specifications

Suitable for Use=Pass
67-56-1
Amber Glass Bottle
6 x 500 mL
100%
0.002-0.030
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit