Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrospec Titrant K5 for Two Component Volumetric Titrations of Aldehydes and Ketones, Ricca Chemical PROMO
Achieve high performance and reliable results. Hydrospec supports a wide range of sample types that help you produce the fast, reliable, and reproducible results you are looking for.
Manufacturer: Ricca Chemical Company RK350000500CPM
Description
- Reliable, Safe, Pyridine-Free
- Available in 500mL and 1L sizes
- 1L Bottles have the Industry Standard GL45 Neck Finish for Convenient Connection
- Stable Formulations and Comprehensive Chemical Compatibility
- 5 Year Shelf Life for Most Reagents
- Technical Support Every Step of the Way
- Tight Specifications for More Accurate Results and Low Lot-to-Lot Variability
- Serving Scientists Successfully for Over 45 Years
Specifications
Suitable for Use=Pass | |
67-56-1, 288-32-4, 7446-09-5 | |
Amber Glass Bottle |
500 mL | |
85%, 10%, 5% | |
0.002-0.030 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title