Learn More
HydroSpec™ Titrant T2 for Two Component Volumetric Titrations with Lower Moisture Content, Ricca Chemical - PROMO
Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.
Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner RK2500006X1CPM
 View more versions of this product
Catalog No. 25-000-061CPM
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
GL45 Neck Finish
7553-56-2, 67-56-1
95, 5
UN1230
0.89
85, 1
Glass Bottle
Liquid