Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrous Benzoyl Peroxide, USP, 70-80%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
1 kg
10 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
94-36-0,7732-18-5
C14H10O4
242.23
OMPJBNCRMGITSC-UHFFFAOYSA-N
benzoyl benzenecarboperoxoate
O=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
94-36-0,7732-18-5
C14H10O4
242.23
OMPJBNCRMGITSC-UHFFFAOYSA-N
benzoyl benzenecarboperoxoate
O=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1

BE156, 94-36-0, C14H10O4 (Anhydrous)

Spectrum™ Hydrous Benzoyl Peroxide, 75 Percent, USP is used to treat mild to moderate acne. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Quantity 12 kg
CAS Max % 0.75,0.25
UN Number UN3104
Percent Purity 70 to 80%