Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrous Benzoyl Peroxide, USP, 70-80%, Spectrum™ Chemical
SDP

Catalog Number 18603279
Click to view available options
Quantity:
25 g
125 g
500 g
1 kg
10 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
94-36-0, 7732-18-5
C14H10O4
242.23
OMPJBNCRMGITSC-UHFFFAOYSA-N
benzoyl benzenecarboperoxoate
O=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
94-36-0, 7732-18-5
C14H10O4
242.23
OMPJBNCRMGITSC-UHFFFAOYSA-N
benzoyl benzenecarboperoxoate
O=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1

BE156, 94-36-0, C14H10O4 (Anhydrous)

Spectrum™ Chemical Hydrous Benzoyl Peroxide, 75 Percent, USP is used to treat mild to moderate acne. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 75%, 25%
Packaging Fiber Drum
Quantity 45 kg
UN Number UN3104
Percent Purity 70 to 80%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit