Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[Hydroxy(methanesulfonyloxy)iodo]benzene 98.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
105551-42-6
C7H9IO4S
316.11
MFCD01321184
WSUYPCBSGVAXNY-UHFFFAOYSA-N
[Hydroxy(mesyloxy)iodo]benzene, Phenyliodosohydroxy Methanesulfonate
11141609
[hydroxy(phenyl)-$l^{3}-iodanyl] methanesulfonate
CS(=O)(=O)O[I](O)C1=CC=CC=C1
105551-42-6
C7H9IO4S
316.11
MFCD01321184
WSUYPCBSGVAXNY-UHFFFAOYSA-N
[Hydroxy(mesyloxy)iodo]benzene, Phenyliodosohydroxy Methanesulfonate
11141609
[hydroxy(phenyl)-$l^{3}-iodanyl] methanesulfonate
CS(=O)(=O)O[I](O)C1=CC=CC=C1
Melting Point 122°C
Color White-Yellow
Quantity 25 g
Formula Weight 316.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [Hydroxy(methanesulfonyloxy)iodo]benzene