Hydroxy Naphthol Blue in NaCl ACS Reagent Grade, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 393025

Packaging Pouch
Quantity 25g

 View more versions of this product

Catalog No. 393025

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 165660-27-5
    CAS Min % 98.0
    CAS Max % 100.0
    Physical Form Powder
    Chemical Name or Material Hydroxy Naphthol Blue, In NaCl
    Grade ACS Reagent Grade

    Specifications

    Physical Form Powder
    Color Gray-purple
    Quantity 25g
    CAS 165660-27-5
    CAS Min % 98.0
    Packaging Pouch
    CAS Max % 100.0
    Chemical Name or Material Hydroxy Naphthol Blue, In NaCl
    Grade ACS Reagent Grade
  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit