Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[Hydroxy(tosyloxy)iodo]benzene, 97+%, Acros Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
27126-76-7
C13H13IO4S
392.21
MFCD00011547
LRIUKPUCKCECPT-UHFFFAOYSA-N
hydroxy tosyloxy iodo benzene,htib,koser's reagent,phenyliodosohydroxy tosylate,koser reagent,unii-2eoa97b14a,hydroxy p-tolylsulfonyl oxy iodo benzene,hydroxy 4-methylbenzenesulfonato-o phenyliodine,iodosobenzene-i-mono-p-toluenesulfonate,hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide
325434
[hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O
27126-76-7
C13H13IO4S
392.21
MFCD00011547
LRIUKPUCKCECPT-UHFFFAOYSA-N
hydroxy tosyloxy iodo benzene,htib,koser's reagent,phenyliodosohydroxy tosylate,koser reagent,unii-2eoa97b14a,hydroxy p-tolylsulfonyl oxy iodo benzene,hydroxy 4-methylbenzenesulfonato-o phenyliodine,iodosobenzene-i-mono-p-toluenesulfonate,hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide
325434
[hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O
oxidation reagent
Packaging Glass bottle
Quantity 25g
Melting Point 131.0°C to 137.0°C
Color Yellow
Infrared Spectrum Authentic
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Merck Index 15, 5369
Formula Weight 392.21
Physical Form Crystalline Powder
Percent Purity 97+%
Chemical Name or Material [Hydroxy(tosyloxy)iodo]benzene
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Warning