Learn More
Alfa Aesar™ Hydroxyapatite Nanoparticles, 5-10% (w/v) aqueous colloidal dispersion
20-50nm particles
Manufacturer:  Alfa Aesar™ J66691AK
 View more versions of this product
Catalog No. AAJ66691AK
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Liquid dispersion
2°C to 8°C
Ca5HO13P3
nHAP; Nano- hydroxyapatite
250mL
12167-74-7
502.31
1100°