Learn More
BioVision IncSupplier Diversity Partner PicoProbe™ β-Hydroxybutyrate Fluorometric Assay Kit: The most sensitive kit on the market to measure (β-HB). Sample type: Serum, plasma, urine or other body fluids, food etc. Detection: < 4 µM. 100 assay DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K651100
β-Hydroxybutyrate (β-HB) is the most abundant of the ketone bodies (~78% of total ketone bodies in blood). Ketoacidosis (high concentrations of the ketone bodies) can be caused by a variety of conditions, such as diabetes, alcoholism and severe starvation. Ketoacidosis can be fatal if not treated. Additionally, ketogenic diets can increase the concentration of ketone bodies slightly in healthy individuals. BioVision’s β-Hydroxybutyrate Assay Kit offers simplicity, enhanced sensitivity, and can be adapted to high-throughput applications. The assay is based on enzymatic oxidation of β-HB that results in generation of fluorescent signal (Ex/Em = 535/587 nm) which is directly proportional to the amount of β-HB. The kit offers an excellent alternative for measurement of β-HB when biological sample quantities are limited or subketogenic levels are suspected. The assay can detect β-Hydroxybutyrate as low as 4 µM in a variety of biological samples. BioVision charges an Ice pack fee on order.
Catalog No. 50-111-9818