Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxychloroquine Sulfate, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
747-36-4
C18H28ClN3O5S
433.95
JCBIVZZPXRZKTI-UHFFFAOYNA-N
2-({4-[(7-chloroquinolin-4-yl)amino]pentyl}(ethyl)amino)ethan-1-ol; sulfuric acid
OS(O)(=O)=O.CCN(CCO)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
747-36-4
C18H28ClN3O5S
433.95
JCBIVZZPXRZKTI-UHFFFAOYNA-N
2-({4-[(7-chloroquinolin-4-yl)amino]pentyl}(ethyl)amino)ethan-1-ol; sulfuric acid
OS(O)(=O)=O.CCN(CCO)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1

H1126, 747-36-4, C18H26ClN3O•H2SO4

Spectrum™ Hydroxychloroquine Sulfate, USP is a medication used for the prevention and treatment of certain types of malaria. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.02
Quantity 5 g
CAS Max % 1
Percent Purity 98 to 102%