Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyethyl Cellulose, 5,000 cP, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9004-62-0
C29H52O21
736.71
CWSZBVAUYPTXTG-UHFFFAOYNA-N
5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol
COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO
9004-62-0
C29H52O21
736.71
CWSZBVAUYPTXTG-UHFFFAOYNA-N
5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol
COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO

H1152, 9004-62-0

Spectrum™ Hydroxyethyl Cellulose, 5,000 cP is also named 2-hydroxyethyl cellulose and is a thickening and gelling agent most commonly used in cleaning solutions, cosmetics and other products used in the household. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Poly Bottle
Loss on Drying 0.05
Quantity 500 g
CAS Max % 1