Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine Hydrochloride, 50% Aqueous Solution, For Mercury, APHA, Spectrum™

Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Amber Glass Bottle
Poly Bottle
5470-11-1
5470-11-1
Packaging Poly Bottle
Quantity 125 mL
Grade Chemical Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.