Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine Hydrochloride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle

Chemical Identifiers

5470-11-1
ClH4NO
69.49
WTDHULULXKLSOZ-UHFFFAOYSA-N
443297
CHEBI:5807
hydroxylamine;hydrochloride
NO.Cl

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 1.67kg
Quantity 500g
Packaging Poly Bottle
Formula Weight 69.49
Physical Form Crystals
Chemical Name or Material Hydroxylamine Hydrochloride
Grade Baker Analyzed™ ACS Reagent