Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydroxylamine Hydrochloride, Crystal, Reagent, ACS, 96%, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Hydroxylamine Hydrochloride, Crystal, Reagent, ACS is a white crystal like substance that is obtained from hydroxylamine. It has many uses, among which some are in organic synthesis, and lignin extraction. Other uses include vulcanization accelerator, stabilizing colors in color film, antioxidant and corrosion inhibitor. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications
CAS Max % | 100% |
Quantity | 6 x 125 g |
UN Number | UN2923 |
Percent Purity | 96% |
Grade | ACS |
Packaging | Amber Glass Bottle |
Residue after Ignition | 0.05% |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction