Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine Hydrochloride, Crystal, Reagent, ACS, 96%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5470-11-1
ClH4NO
69.49
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydrogen hydroxylamine chloride
[H+].[Cl-].NO
5470-11-1
ClH4NO
69.49
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydrogen hydroxylamine chloride
[H+].[Cl-].NO

H1085, 5470-11-1, NH2OH•HCl

Spectrum™ Hydroxylamine Hydrochloride, Crystal, Reagent, ACS is a white crystal like substance that is obtained from hydroxylamine. It has many uses, among which some are in organic synthesis, and lignin extraction. Other uses include vulcanization accelerator, stabilizing colors in color film, antioxidant and corrosion inhibitor. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
CAS Max % 1
UN Number UN2923
Percent Purity 96%
Other Heavy Metals 5 ppm
Packaging Amber Glass Bottle
Residue after Ignition 0.0005
Sulfur (S) 5e-005
Quantity 500 g