Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine sulfate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
5kg
Packaging:
Glass Bottle
Plastic bottle
10039-54-0
H6N2O2·H2SO4
MFCD00044869
Hydroxylammonium sulfate
10039-54-0
H6N2O2·H2SO4
MFCD00044869
Hydroxylammonium sulfate
Infrared Spectrum Authentic
Packaging Glass Bottle
Color White
Quantity 5g
Assay Percent Range 99%
Linear Formula (H2NOH)2·H2SO4
Fieser 01,481
Merck Index 15, 4867
Formula Weight 164.14
Physical Form Crystals
Percent Purity ≥98.5%
Chemical Name or Material Hydroxylamine sulfate
Show More Show Less

GHS P Statement
May be corrosive to metals.
Suspected of causing cancer if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger