Hydroxylamine sulfate, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 8894414

Quantity 25g

 View more versions of this product

Catalog No. AA8894414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10039-54-0
    Physical Form Crystalline
    Assay Percent Range 99%
    Molecular Formula H3NO·0.5H2SO4
    Formula Weight 164.14
    MDL Number MFCD00044869
    Chemical Name or Material Hydroxylamine sulfate

    Specifications

    Formula Weight 164.14
    Quantity 25g
    EINECS Number 233-118-8
    Physical Form Crystalline
    Assay Percent Range 99%
    Molecular Formula H3NO·0.5H2SO4
    MDL Number MFCD00044869
    CAS 10039-54-0
    Chemical Name or Material Hydroxylamine sulfate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit