Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine Sulfate, BAKER™, J.T.Baker™

Click to view available options
Quantity:
250g
500g
Packaging:
Poly Bottle

Chemical Identifiers

10039-54-0
(NH2OH)2·H2SO4

A grade of chemicals for laboratory use

Specific Gravity 1L = 1.90kg
Quantity 500g
Packaging Poly Bottle
Formula Weight 164.14
Chemical Name or Material Hydroxylamine Sulfate