Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine Sulfate, BAKER™, J.T. Baker™

Catalog No. p-5391210
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 10039-54-0
Molecular Formula (NH2OH)2·H2SO4

Specifications

Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 1.90kg
Formula Weight 164.14
Chemical Name or Material Hydroxylamine Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.