Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine Sulfate, Crystal, 98%, Spectrum™

Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10039-54-0
10039-54-0
Packaging Poly Pail
Residue after Ignition 0.0005
Quantity 2.5 kg
CAS Max % 1
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.