Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylapatite, for analysis, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Glass bottle
Plastic Bottle
1306-06-5
HCa5O13P3
502.31
MFCD00010904
Calcium phosphate hydroxide
1306-06-5
HCa5O13P3
502.31
MFCD00010904
Calcium phosphate hydroxide
Linear Formula 3Ca3(PO4)2·Ca(OH)2
Merck Index 15, 3514
Arsenic (As) 2ppm max.
Solubility Information Solubility in water: negligible
Formula Weight 502.31
Physical Form Powder
Percent Purity 35 to 40% (Ca)
Grade Analysis
Chloride 1500ppm max
Chlorine (Cl) 1500ppm max.
Copper (Cu) 20ppm max.
Fluorine (F) 50ppm max.
Heavy Metals (as Pb) 10ppm max.
Insoluble in Water 0.2% max. (in HCl)
Insoluble Matter 0.2% max. (in HCl)
Iron (Fe) 400ppm max.
Lead (Pb) 5ppm max.
Loss on Ignition 8% max. (800°C)
Mercury (Hg) 1ppm max.
Packaging Glass bottle
Solubility Solubility in water: negligible.
Sulfate 5000ppm max.
Trace Analysis 20ppm max.
Color White
Melting Point 1700.0°C
Quantity 25g
Chemical Name or Material Hydroxylapatite
Show More Show Less

EINECSNumber : 215-145-7