Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxymethylpolystyrene resin, 1% crossl. with DVB,0.4-0.7mmol/g, 200-400 mesh, Acros Organics

Click to view available options
Quantity:
5g
25g
66072-40-0
C7H8O
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
Benzylalcohol polymer bound
244
CHEBI:17987
phenylmethanol
C1=CC=C(C=C1)CO
66072-40-0
C7H8O
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
Benzylalcohol polymer bound
244
CHEBI:17987
phenylmethanol
C1=CC=C(C=C1)CO
for solid support synthesis
Additional Information crosslinking: 1% DVB
Loading 0.4 to 0.7mmol/g
Mesh Size 200 to 400 mesh
Quantity 25g
Color White
Physical Form Beads
Chemical Name or Material Hydroxymethylpolystyrene resin