Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyprogesterone Caproate, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
630-56-8
C27H40O4
428.61
DOMWKUIIPQCAJU-LJHIYBGHSA-N
(1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl hexanoate
CCCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
630-56-8
C27H40O4
428.61
DOMWKUIIPQCAJU-LJHIYBGHSA-N
(1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl hexanoate
CCCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O

H1234, 630-56-8, C27H40O4

Spectrum™ Hydroxyprogesterone Caproate, USP is a prescription hormone medication used to lower the risk of preterm birth in women who are pregnant with one baby and who have delivered one baby too early (preterm) in the past. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Melting Range 120°-124°C
Specific Rotation +58° to +64°
Quantity 500 g
CAS Max % 1
Percent Purity 97 to 103%