Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxypropyl Cellulose, 150-400 cP, NF, 53.4-850%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9004-64-2
(C24H44O16)n
NaN
CC(O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)O)OC(-*)[C@H](OC(C)O)[C@H]2OC(C)O)[C@H](OC(C)O)[C@@H](OC(C)O)[C@@H]1O-*
9004-64-2
(C24H44O16)n
NaN
CC(O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)O)OC(-*)[C@H](OC(C)O)[C@H]2OC(C)O)[C@H](OC(C)O)[C@@H](OC(C)O)[C@@H]1O-*

HY119, 9004-64-2

Spectrum™ Hydroxypropyl Cellulose, 150-400 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Ignition Residue 0.008
Loss on Drying 0.05
Quantity 500 g
CAS Max % 1
Percent Purity 53.4 to 80.5%