Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyurea, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
127-07-1
CH4N2O2
76.04
MFCD00007943
VSNHCAURESNICA-UHFFFAOYSA-N
hydroxycarbamide,n-hydroxyurea,hydrea,1-hydroxyurea,oxyurea,carbamoyl oxime,biosupressin,hydroxycarbamine,onco-carbide,carbamohydroxamic acid
3657
CHEBI:44423
hydroxyurea
C(=O)(N)NO
127-07-1
CH4N2O2
76.04
MFCD00007943
VSNHCAURESNICA-UHFFFAOYSA-N
hydroxycarbamide,n-hydroxyurea,hydrea,1-hydroxyurea,oxyurea,carbamoyl oxime,biosupressin,hydroxycarbamine,onco-carbide,carbamohydroxamic acid
3657
CHEBI:44423
hydroxyurea
C(=O)(N)NO
Melting Point 142°C to 146°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Loss on Drying 1% max. (1 g, 60°C) (< 0.7 kPa)
Color White
Assay Percent Range 98%
Linear Formula HONHCONH2
Beilstein 03, 95
Merck Index 15, 4888
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in hot alcohol
Formula Weight 76.04
Physical Form Crystalline Powder
Percent Purity ≥97.5% (on dried substance)
Chemical Name or Material Hydroxyurea
Show More Show Less

GHS H Statement:
May cause genetic defects.
May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.