Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyurea, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
50 g
100 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
127-07-1
CH4N2O2
76.06
VSNHCAURESNICA-UHFFFAOYSA-N
hydroxyurea
NC(=O)NO
127-07-1
CH4N2O2
76.06
VSNHCAURESNICA-UHFFFAOYSA-N
hydroxyurea
NC(=O)NO

H1181, 127-07-1, CH4N2O2

Spectrum™ Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.005
Loss on Drying 0.01
Quantity 100 g
CAS Max % 1
Percent Purity 97 to 103%