Learn More
Ansell™ HyFlex™ 11-281 Sleeves
Ansell™ HyFlex™ 11-281 Sleeves feature a seamless design and options of lengths and widths to ensure an excellent fit for all-day comfort
$483.00 - $621.50
Specifications
Certifications/Compliance REACH Compliant
Certifications/Compliance (Europe) EN420:2003 + A1:2009; Category ll
Cleaning Procedure 40°C (104°F)
Color Gray
Construction Knitted
View More Specs
Products 8
Catalog Number Mfr. No. Size Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size Length (English) Price Quantity & Availability  
19-500-741 View Documents Ansell™
11281120N
Narrow 12 in. Case of 50 for $483.00
Add to cart
19-500-742 View Documents Ansell™
11281120W
Wide 12 in. Case of 50 for $483.00
Add to cart
19-500-743 View Documents Ansell™
11281160N
Narrow 16 in. Case of 50 for $558.50
Add to cart
19-500-744 View Documents Ansell™
11281160W
Wide 16 in. Case of 50 for $558.50
Add to cart
19-500-745 View Documents Ansell™
11281180N
Narrow 18 in. Case of 50 for $596.50
Add to cart
19-500-746 View Documents Ansell™
11281180W
Wide 18 in. Case of 50 for $596.50
Add to cart
19-500-747 View Documents Ansell™
11281220N
Narrow 22 in. Case of 50 for $621.50
Add to cart
19-500-748 View Documents Ansell™
11281220W
Wide 22 in. Case of 50 for $621.50
Add to cart
Description
Description
  • INTERCEPT™ Cut Resistance Technology provides best-in-class cut protection and a soft and cool feel to the worker
  • Engineered to perform in combination with any HyFlex cut resistant glove
  • Available in three versions: Extra Wide, Wide, Narrow
Specifications
Specifications
REACH Compliant
40°C (104°F)
Knitted
No
ANSELL INTERCEPT™ Technology
Non-sterile
15 Gauge
EN420:2003 + A1:2009; Category ll
Gray
A4
Glass repair and replacement; Handling metal sheets and panels
Sleeve
HyFlex™11-281