Learn More
Ansell™ HyFlex™ Light-Duty Multipurpose Industrial Gloves
Designed for light and delicate applications in dry environments that require maximal levels of dexterity. Like a second skin, they offer comfortable, exact handling of small objects. They also ensure product protection from glove-generated debris, and cross-product contamination.
$72.10
Specifications
Glove Cuff Style Knit
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-152-159
View Documents
Ansell™
205653
9 Dozen of 12 for $72.10
Case of 12 DZ for $756.00
Add to cart
Description
Description
  • Industrial gloves
  • Offer comfortable, exact handling of small objects
  • Ensures product protection from glove-generated debris, and cross-product contamination
  • Provides precision and/or clean handling
Applications:
  • Assembly of small pieces such as small bearings, joints and springs
Compliance
Meets ANSI A1 (cut level) and ANSI 3 (abrasion level); EN 388-3121 and EN 1149 standards.
Specifications
Specifications
Knit