Learn More
Ansell™ HyFlex™ 11-605 Series White Polyurethane Light Weight Gloves
Designed for light and delicate applications in dry environments requiring maximum levels of dexterity
Specifications
Back Material Polyurethane
Certifications/Compliance Standards CATEGORY III, EN 388 : 003X, ANSI, CE Mark
Coating Material Polyurethane
Cuff Material Polyurethane
Glove Cuff Style Knitwrist
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-133-483
View Documents
Ansell™
11605 6 286151
6 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-042-611
View Documents
Ansell™
11605 7 286152
7 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Polyurethene construction
  • Knitted gloves
  • Nylon liner
  • Fingertip-coated finishing
  • Knitwrist cuff
Specifications
Specifications
Polyurethane
Polyurethane
Knitwrist
Coated-palm
11-605
Standards CATEGORY III, EN 388 : 003X, ANSI, CE Mark
Polyurethane
Nylon
Precision, cut-resistant, oil-repellant, and specialty products