Learn More
Ansell™ HyFlex™ 11-627 Series Gray Polyurethane Medium Weight Gloves
Offers high levels of cut resistance for a safe use in controlling, handling and assembly applications involving contacts with sharp pieces. The Lycra™ in the liners of the gloves as well as in the polyurethane coating provides excellent flexibility and fit.
Manufacturer:  Ansell™ 205688
 View more versions of this product
Catalog No. 19-168-52
Add to cart
Description
Description
  • Industrial gloves
  • Special formulation of the polyurethane coating gives the gloves flexibility and fit, as well as secure grip on dry to slightly oily pieces for sure handling in wide range of environments
  • Unique combination of Dyneema™, Nylon, Lycra™, and glass fiber
  • Ultra strong fibers provide extreme resistance to cuts and burrs
  • Delivers perfect balance of comfort and sure handling with superior cut resistance
  • For safe use in controlling, handling and assembly applications involving contacts with sharp pieces
  • Lycra™ in the liners of these gloves, as well as the polyurethane coating, provides excellent flexibility and fit
Applications:
  • Assembling and mounting components
  • Assembly & inspection of components
  • Cutting glasses, sheets, panels
  • Handling materials with sharp or rough edges
  • Handling parts with sharp, rough edges
  • Handling rough machined metal parts
  • Picking, fastening components
  • Press injection molding
Specifications
Specifications
Polyurethane
Knit
Plastics injection and molding, Glass handling, sheet Metal assembly (even when slightly Oily), handling sharp, small parts
8
Gray
Level 2
Yellow
Dyneema™, Lycra™
Medium weight