Learn More
Ansell™ HyFlex™ 11-627 Series Gray Polyurethane Medium Weight Gloves
Hyflex™ CR2 Gloves offer high levels of cut resistance for a safe use in controlling, handling and assembly applications involving contacts with sharp pieces. The Lycra™ in the liners of the gloves as well as in the polyurethane coating provides excellent flexibility and fit.
$340.00
Specifications
Glove Cuff Style Knit
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-168-50
View Documents
Ansell™
205686
6
Dozen of 12 for $340.00
Case of 12 DZ for $3,590.00
Add to cart
19-168-51
View Documents
Ansell™
205687
7
Dozen of 12 for $340.00
Case of 12 DZ for $3,579.48
Add to cart
19-168-52
View Documents
Ansell™
205688
8
Dozen of 12 for $340.00
Case of 12 DZ for $3,579.48
Add to cart
19-168-53
View Documents
Ansell™
205689
9
Dozen of 12 for $340.00
Case of 12 DZ for $3,579.48
Add to cart
19-168-54
View Documents
Ansell™
205690
10
Dozen of 12 for $340.00
Case of 12 DZ for $3,579.48
Add to cart
19-168-55
View Documents
Ansell™
205709
11
Dozen of 12 for $340.00
Case of 12 DZ for $3,579.48
Add to cart
Description
Description
  • Offer excellent abrasion resistance, cut protection, dexterity and comfort
  • Polyurethane dipped: Special formulation of the polyurethane coating gives the gloves flexibility and fit, as well as secure grip on dry to slightly oily pieces for sure handling in wide range of environments
  • Unique combination of Dyneema™, Nylon, Lycra™, and glass fiber
  • Ultra strong fibers provide extreme resistance to cuts and burrs
  • Delivers perfect balance of comfort and sure handling with superior cut resistance
  • For safe use in controlling, handling and assembly applications involving contacts with sharp pieces
  • Palm coating provides dry grip and longer wear
  • Natural fit, Dyneema™ and Lycra™ liner reduces hand fatigue
  • Excellent grip in dry environments; effective control in minimally oily environments
  • Breathable liner increases worker acceptance and safety compliance
  • Machine launderable
  • ANSI™ Level 2 cut protection
Applications
:
  • Assembling and mounting components
  • Assembly & inspection of components
  • Cutting glasses, sheets, panels
  • Handling materials with sharp or rough edges
  • Handling parts with sharp, rough edges
  • Handling rough machined metal parts
  • Picking, fastening components
  • Press injection molding
Specifications
Specifications
Knit