Learn More
Ansell™ HyFlex™ 47-400 Series Blue Nitrile Medium Weight Gloves
Versatile alternative to cotton, light leather and PVC. Special nitrile formulation provides an excellent combination of flexibility, mechanical resistance and oil repellency.
$22.05 - $104.00
Save up to 78%
Specifications
Glove Cuff Style Knit
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-019867
View Documents
Ansell™
47400 10 205934
10
Dozen of 12 for $22.05
Save 78% Reg : $101.00
Case of 12 DZ for $264.60
Save 76% Reg : $1,081.48
Add to cart
19-019860
View Documents
Ansell™
47400 8 205931
8
Dozen of 12 for $104.00
Case of 12 DZ for $1,107.08
Add to cart
19-019865
View Documents
Ansell™
205933
9
Dozen of 12 for $103.70
Case of 12 DZ for $1,113.00
Add to cart
Description
Description
Ideal for assembly, inspection, light fabrication, materials handling, shipping and receiving, maintenance, and sheet metal work.
  • Natural hand shape - easier to wear for long periods.
  • Silicone-free.
  • Ensures cleanliness of metal sheets: important in steel and motor industries. 
  • Cotton interlock lining with knitwrist for extra comfort
  • Nitrile-coated palm for exceptional cut
  • Offer puncture and abrasion resistance
  • Provide flexibility and dexterity
  • Machine washable; can be laundered repeatedly without becoming stiff and uncomfortable
Specifications
Specifications
Knit