Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ AlphaTec™ Butyl Glove 38-514

The thinnest butyl glove providing the best resistance to the most aggressive chemicals with dexterity and comfort.

Manufacturer:  Ansell™ 189024

 View more versions of this product

Catalog No. 19-014-453



Add to cart
This item is not returnable. View return policy

Description

Description

Provides the best resistance to the most aggressive chemicals without compromising dexterity or comfort. Offers superior protection against aldehydes, ketones and esters as well as concentrated mineral acids.

  • 38-514 offers superior protection against aldehydes, ketones and esters as well as concentrated mineral acids
  • The glove is very flexible and provides excellent dexterity
  • Designed for comfort, with a natural, curved ergonomic shape and soft feel, they offer easy donning and good grip
  • The absence of latex proteins means there’s no risk of latex allergies
  • Rough finish on the hand section of the glove for safer handling
Specifications

Specifications

Black
14 in.
36 pairs in a carton; individually wrapped with insert
EN 420:2003 + A1:2009, CE 0493
EN388:1000X
ABCIKMPST
No
Beaded
350 mm
10
ANSI 3
Type A
Butyl Polymer
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit