Learn More
Hygiena™ MicroSnap Coliform Rapid Coliform Detection
Rapid test for detection and enumeration of Coliform bacteria
Manufacturer:  HYGIENA LLC MSCC100
 View more versions of this product
Catalog No. 14-037-783
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Coliform Test Indicator
Hygiena EnSURE™ luminometer
Luciferase Enzyme, Synthetic D-luciferin ((S)-4, 5-dihydro-2(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic Acid), proprietary Substrates, stabilizers, Buffers, Sodium Azide and detergent, proprietary lysing agent in tube
2° to 8°C